Läs mer
enQuireBio™ Recombinant M. tuberculosis Immunogenic protein MPT63 Protein
Produktkod. p-7155447
Kvantitet:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Förpackningsstorlek:
100µg
10µg
1mg
200µg
500µg
50µg
Produktkod. 16016661
Brand: enQuireBio™ QP8820ec10ug
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
A DNA sequence encoding the Mycobacterium tuberculosis (strain CDC 1551 / Oshkosh) Immunogenic protein MPT63, was expressed in the hosts and tags indicated.
Specifikationer
Tillträdesnummer WP_003409684.1
Namn M. tuberculosis Immunogenic protein MPT63 Protein
Kvantitet 10 μg
Regulatorisk status Research Use Only
Endotoxinkoncentration Not determined.
Gensymbol mpt63
Protein Längd Full Length
Produkttyp Recombinant Protein
Korsreaktivitet M. tuberculosis
Art E. coli
Protein Tag His
Sekvens AYPITGKLGS ELTMTDTVGQ VVLGWKVSDL KSSTAVIPGY PVAGQVWEAT ATVNAIRGSV TPAVSQFNAR TADGINYRVL WQAAGPDTIS GATIPQGEQS TGKIYFDVTG PSPTIVAMNN GMEDLLIWEP
Buffert Tris-based buffer, 50% glycerol
Renhet eller kvalitetsklass Greater than 90% as determined by SDS-PAGE.