missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Macaque IL-15-S48AA Protein

Product Code. 16057851
1 mg, 1mg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy
This item is not returnable. View return policy

A DNA sequence encoding the Macaca mulatta (Rhesus macaque) IL-15-S48AA, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001038196.1
Gene ID (Entrez) 699616
Name IL-15-S48AA Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol IL15
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Macaque
Species Yeast
Protein Tag His
Sequence NWVNVISDLK KIEDLIQSMH IDATLYTESD VHPSCKVTAM KCFLLELQVI SHESGDTDIH DTVENLIILA NNILSSNGNI TESGCKECEE LEEKNIKEFL QSFVHIVQMF INTS
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Macaque IL-15-S48AA Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.