missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Adiponectin Protein

A DNA sequence encoding the Mus musculus (Mouse) Adiponectin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8593-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16055471

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_033735.3
Adiponectin Protein
500 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
11450
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ma/Adipoq
Recombinant Protein
E. coli
EDDVTTTEEL APALVPPPKG TCAGWMAGIP GHPGHNGTPG RDGRDGTPGE KGEKGDAGLL GPKGETGDVG MTGAEGPRGF PGTPGRKGEP GEAAYVYRSA FSVGLETRVT VPNVPIRFTK IFYNQQNHYD GSTGKFYCNI PGLYYFSYHI TVYMKDVKVS LFKKDKAVLF TYDQYQEKNV DQASGSVLLH LEVGDQVWLQ VYGDGDHNGL YADNVNDSTF TGFLLYHDTN
Videos
SDS
Documents