missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse b NGF Protein
A cDNA sequence encoding the b NGF was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10528-20ug
Additional Details : Weight : 0.01000kg
Specifications
b NGF Protein | |
Research Use Only | |
The method used to test the bioassay is the NGF-dependent survival of dorsal root ganglia neurons of chick embryo, corresponding to a Specific Activity of 500,000IU/mg. | |
Mouse | |
Untagged | |
The NGF beta Mouse was lyophilized from solution containing 5% mannitol and 1% HSA. |
20 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
SSTHPVFHMGEF SVCDSVSVWV GDKTTATDIK GKEVTVLAEV NINNSVFRQY FFETKCRASN PVESGCRGID SKHWNSYCTT THTFVKALTT DEKQAAWRFI RIDTACVCVL SRKATRRG | |
Greater than 98% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE . |