missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse C-C chemokine receptor type 5 Protein

A DNA sequence encoding the Mus musculus (Mouse) C-C chemokine receptor type 5, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9040-ye-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16047501

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_034047.2
C-C chemokine receptor type 5 Protein
Research Use Only
Ccr5
Recombinant Protein
Yeast
QEFFGLNNCS SSNRLDQAMQ ATETLGMTHC CLNPVIYAFV GEKFRSYLSV FFRKHMVKRF CKRCSIFQQD NPDRASSVYT RSTGEHEVST GL
Greater than 90% as determined by SDS-PAGE.
12774
10 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers