Läs mer
R&D Systems™ Recombinant Mouse CCL1/TCA-3 Protein
Formulering:
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Beskrivning
Measured by its ability to chemoattract BaF3 mouse pro-B cells transfected with mouse CCR8. The ED50 for this effect is 0.5-2.0 ng/mL.
Specifikationer
Specifikationer
| Formulering | Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. |
| Gen-ID (Entrez) | 20290 |
| Molekylvikt (g/mol) | 7.8 kDa |
| Namn | CCL1/I-309/TCA-3 |
| Kvantitet | 25 μg |
| Källa | Mouse myeloma cell line,NS0-derived mouse CCL1/I-309/TCA-3 protein Lys24-Cys92 |
| Förvaringskrav | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Konjugera | Unconjugated |
| Rekombinant | Recombinant |
| Renhet eller kvalitetsklass | >95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |