Läs mer
Novus Biologicals™ Recombinant Mouse CCL25/TECK Protein
Produktkod. 18437541 Handla allt Bio Techne Produkter
Kvantitet:
100 μg
1mg
5 μg
500 μg
Produktkod. 18437541
Brand: Novus Biologicals™ NBP235184500ug
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Recombinant Protein
CCL25/TECK protein is fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using human monocytes is in a concentration range of 5.0-50 ng/mL
TRUSTED_SUSTAINABILITY
Specifikationer
Tillträdesnummer O35903, O35903, O35903
För användning med (applikation) PAGE, Bioactivity
Formulering Lyophilized from a 0.2 um filtered concentrated solution in 20 mM PB, pH 7.4, 150 mM NaCl.
Gen-ID (Entrez) 6370
Molekylvikt (g/mol) M.W. (Theoretical): 14.1 kDa
Kvantitet 500 μg
Förvaringskrav Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxinkoncentration Less than 1 EU/μg of CCL25/TECK as determined by LAL method.
Gene Alias C-C motif chemokine 25, CCL25, chemokine (C-C motif) ligand 25, Chemokine TECK, Ck beta-15, Ckb15, SCYA25TECKvar, small inducible cytokine subfamily A (Cys-Cys), member 25, Small-inducible cytokine A25, TECK/CCL25, TECKMGC150327, thymus expressed chemokine, Thymus-expressed chemokine
Gensymbol CCL25
Rekonstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C
Forskningskategori Biologically Active Proteins, Chemokines and Cytokines, Signal Transduction
Renhet eller kvalitetsklass >95%, by SDS-PAGE and HPLC
Protein CCL25/TECK