missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CD90 / THY-1 Protein

A DNA sequence encoding the Mus musculus (Mouse) CD90 / THY-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6789-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16065870

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_033408.1
CD90 / THY-1 Protein
200 μg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
21838
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Thy1
Recombinant Protein
E. coli
QKVTSLTACL VNQNLRLDCR HENNTKDNSI QHEFSLTREK RKHVLSGTLG IPEHTYRSRV TLSNQPYIKV LTLANFTTKD EGDYFCELQV SGANPMSSNK SISVYRDKLV KC
Videos
SDS
Documents