missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CLEC4E / CLECSF9 Protein

A DNA sequence encoding the Mus musculus (Mouse) CLEC4E / CLECSF9, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8099-ec-10ug

 View more versions of this product

Product Code. 16032671

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_064332.1
CLEC4E / CLECSF9 Protein
Research Use Only
Clec4e
Recombinant Protein
E. coli
TYRSSQISGQ NLQPHRNIKE LSCYSEASGS VKNCCPLNWK HYQSSCYFFS TTTLTWSSSL KNCSDMGAHL VVIDTQEEQE FLFRTKPKRK EFYIGLTDQV VEGQWQWVDD TPFTESLSFW DAGEPNNIVL VEDCATIRDS SNSRKNWNDI PCFYSMPWIC EMPEISPLD
Greater than 90% as determined by SDS-PAGE.
56619
10 μg
Not determined.
Extracellular Domain
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers