missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CLEC4E / CLECSF9 Protein
A DNA sequence encoding the Mus musculus (Mouse) CLEC4E / CLECSF9, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8099-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_064332.1 | |
CLEC4E / CLECSF9 Protein | |
Research Use Only | |
Clec4e | |
Recombinant Protein | |
E. coli | |
TYRSSQISGQ NLQPHRNIKE LSCYSEASGS VKNCCPLNWK HYQSSCYFFS TTTLTWSSSL KNCSDMGAHL VVIDTQEEQE FLFRTKPKRK EFYIGLTDQV VEGQWQWVDD TPFTESLSFW DAGEPNNIVL VEDCATIRDS SNSRKNWNDI PCFYSMPWIC EMPEISPLD | |
Greater than 90% as determined by SDS-PAGE. |
56619 | |
500 μg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |