Läs mer
R&D Systems™ Recombinant Mouse Coagulation Factor Xa Protein
Kvantitet:
10 μg
Beskrivning
The Recombinant Mouse Coagulation Factor Xa Protein is derived from Sf 21 (baculovirus). The Recombinant Mouse Coagulation Factor Xa Protein has been validated for the following applications: Enzyme Activity.
Specifikationer
Specifikationer
| Formulering | Supplied as a 0.2 μm filtered solution in MES, NaCl and CaCl 2 . |
| Gen-ID (Entrez) | 14058 |
| Molekylvikt (g/mol) | 29 kDa & 10 kDa |
| Namn | Coagulation Factor X |
| Kvantitet | 10 μg |
| Källa | Spodoptera frugiperda,Sf 21 (baculovirus)-derived mouse Coagulation Factor X protein Met1-Asn481,with a C-terminal 10-His tag. Accession &Num; O88947 Proform Factor X was expressed,purified,activated and further purified to yield Factor Xa. |
| Förvaringskrav | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Strukturell form | Recombinant Mouse Coagulation Factor X is prone to proteolytic cleavage at C-terminus. The predominant form of the purified protein lacks the His tag. |
| Konjugera | Unconjugated |
| Rekombinant | Recombinant |
| Visa mer |