missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CXCL2 / MIP-2 Protein

A DNA sequence encoding the Mus musculus (Mouse) CXCL2 / MIP-2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8651-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16055771

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_033166.1
CXCL2 / MIP-2 Protein
200 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
20310
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CXCL2
Recombinant Protein
E. coli
AVVASELRCQ CLKTLPRVDF KNIQSLSVTP PGPHCAQTEV IATLKGGQKV CLDPEAPLVQ KIIQKILNKG KAN
Videos
SDS
Documents