missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CXCL2 / MIP-2 Protein
A DNA sequence encoding the Mus musculus (Mouse) CXCL2 / MIP-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8651-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_033166.1 | |
CXCL2 / MIP-2 Protein | |
Research Use Only | |
CXCL2 | |
Recombinant Protein | |
E. coli | |
AVVASELRCQ CLKTLPRVDF KNIQSLSVTP PGPHCAQTEV IATLKGGQKV CLDPEAPLVQ KIIQKILNKG KAN | |
Greater than 90% as determined by SDS-PAGE. |
20310 | |
500 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |