missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Galectin-4 Protein
A DNA sequence encoding the Mus musculus (Mouse) Galectin-4, was expressed in the hosts and tags indicated.
2955.00 SEK - 46145.00 SEK
Specifications
Accession Number | NP_034836.1 |
---|---|
Gene ID (Entrez) | 16855 |
Name | Galectin-4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16038841
|
enQuireBio™
QP9839-YE-100UG |
100 μg |
10410.00 SEK
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16058841
|
enQuireBio™
QP9839-YE-1MG |
1 mg |
46145.00 SEK
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16048841
|
enQuireBio™
QP9839-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16068841
|
enQuireBio™
QP9839-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078841
|
enQuireBio™
QP9839-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16088841
|
enQuireBio™
QP9839-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_034836.1 | |
Galectin-4 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
16855 | |
Research Use Only | |
Lgals4 | |
Recombinant Protein | |
Yeast | |
MAYVPAPGYQ PTYNPTLPYK RPIPGGLSVG MSVYIQGMAK ENMRRFHVNF AVGQDDGADV AFHFNPRFDG WDKVVFNTMQ SGQWGKEEKK KSMPFQKGKH FELVFMVMPE HYKVVVNGNS FYEYGHRLPV QMVTHLQVDG DLELQSINFL GGQPAAAPYP GAMTIPAYPA GSPGYNPPQM NTLPVMTGPP VFNPRVPYVG ALQGGLTVRR TIIIKGYVLP TARNFVINFK VGSSGDIALH LNPRIGDSVV RNSFMNGSWG AEERKVAYNP FGPGQFFDLS IRCGMDRFKV FANGQHLFDF SHRFQAFQMV DTLEINGDIT LSYVQI | |
Greater than 90% as determined by SDS-PAGE. |