missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Galectin-4 Protein

A DNA sequence encoding the Mus musculus (Mouse) Galectin-4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9839-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16058841

  • 46145.00 SEK / 1mg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_034836.1
Galectin-4 Protein
Research Use Only
Lgals4
Recombinant Protein
Yeast
MAYVPAPGYQ PTYNPTLPYK RPIPGGLSVG MSVYIQGMAK ENMRRFHVNF AVGQDDGADV AFHFNPRFDG WDKVVFNTMQ SGQWGKEEKK KSMPFQKGKH FELVFMVMPE HYKVVVNGNS FYEYGHRLPV QMVTHLQVDG DLELQSINFL GGQPAAAPYP GAMTIPAYPA GSPGYNPPQM NTLPVMTGPP VFNPRVPYVG ALQGGLTVRR TIIIKGYVLP TARNFVINFK VGSSGDIALH LNPRIGDSVV RNSFMNGSWG AEERKVAYNP FGPGQFFDLS IRCGMDRFKV FANGQHLFDF SHRFQAFQMV DTLEINGDIT LSYVQI
Greater than 90% as determined by SDS-PAGE.
16855
1 mg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers