missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Glucose-6-phosphate isomerase Protein
A DNA sequence encoding the Mus musculus (Mouse) Glucose-6-phosphate isomerase, was expressed in the hosts and tags indicated.
2430.00 SEK - 27205.00 SEK
Specifications
Accession Number | NP_032181.2 |
---|---|
Gene ID (Entrez) | 14751 |
Name | Glucose-6-phosphate isomerase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16047751
|
enQuireBio™
QP9161-YE-100UG |
100 μg |
8695.00 SEK
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16087751
|
enQuireBio™
QP9161-YE-500UG |
500 μg |
24845.00 SEK
500µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16057751
|
enQuireBio™
QP9161-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16077751
|
enQuireBio™
QP9161-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16097751
|
enQuireBio™
QP9161-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16067751
|
enQuireBio™
QP9161-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_032181.2 | |
Glucose-6-phosphate isomerase Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
14751 | |
Research Use Only | |
Gpi | |
Recombinant Protein | |
Yeast | |
AALTRNPQFQ KLLEWHRANS ANLKLRELFE ADPERFNNFS LNLNTNHGHI LVDYSKNLVN KEVMQMLVEL AKSRGVEAAR DNMFSGSKIN YTENRAVLHV ALRNRSNTPI KVDGKDVMPE VNRVLDKMKS FCQRVRSGDW KGYTGKSITD IINIGIGGSD LGPLMVTEAL KPYSKGGPRV WFVSNIDGTH IAKTLASLSP ETSLFIIASK TFTTQETITN AETAKEWFLE AAKDPSAVAK HFVALSTNTA KVKEFGIDPQ NMFEFWDWVG GRYSLWSAIG LSIALHVGFD HFEQLLSGAH WMDQHFLKTP LEKNAPVLLA LLGIWYINCY GCETHALLPY DQYMHRFAAY FQQGDMESNG KYITKSGARV DHQTGPIVWG EPGTNGQHAF YQLIHQGTKM IPCDFLIPVQ TQHPIRKGLH HKILLANFLA QTEALMKGKL PEEARKELQA AGKSPEDLEK LLPHKVFEGN RPTNSIVFTK LTPFILGALI AMYEHKIFVQ GIMWDINSFD QWGVELGKQL AKKIEPELEG SSAVTSHDSS TNGLISFIKQ QRDTKLE | |
Greater than 90% as determined by SDS-PAGE. |