Läs mer
enQuireBio™ Recombinant Mouse Hemoglobin subunit alpha Protein
Produktkod. 16061160
50 μg, 50µg
Kvantitet:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Förpackningsstorlek:
100µg
10µg
1mg
200µg
500µg
50µg
Produktkod. 16061160
Brand: enQuireBio™ QP6146ye50ug
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
A DNA sequence encoding the Mus musculus (Mouse) Hemoglobin subunit alpha, was expressed in the hosts and tags indicated.
Specifikationer
Namn Hemoglobin subunit alpha Protein
Kvantitet 50 μg
Regulatorisk status Research Use Only
Endotoxinkoncentration Not determined.
Gensymbol Hba
Protein Längd Full Length
Produkttyp Recombinant Protein
Korsreaktivitet Mouse
Art Yeast
Protein Tag His
Sekvens VLSGEDKSNI KAAWGKIGGH GAEYGAEALE RMFASFPTTK TYFPHFDVSH GSAQVKGHGK KVADALASAA GHLDDLPGAL SALSDLHAHK LRVDPVNFKL LSHCLLVTLA SHHPADFTPA VHASLDKFLA SVSTVLTSKY R
Buffert Tris-based buffer, 50% glycerol
Renhet eller kvalitetsklass Greater than 80% as determined by SDS-PAGE.
Produkttitel
enQuireBio™ Recombinant Mouse Hemoglobin subunit alpha Protein >