missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Hemoglobin subunit beta-2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Hemoglobin subunit beta-2, was expressed in the hosts and tags indicated.
2430.00 SEK - 26260.00 SEK
Specifications
Accession Number | NP_058652.1 |
---|---|
Gene ID (Entrez) | 15130 |
Name | Hemoglobin subunit beta-2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16028360
|
enQuireBio™
QP7346-YE-100UG |
100 μg |
8695.00 SEK
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16068360
|
enQuireBio™
QP7346-YE-500UG |
500 μg |
24845.00 SEK
500µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16038360
|
enQuireBio™
QP7346-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058360
|
enQuireBio™
QP7346-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078360
|
enQuireBio™
QP7346-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16048360
|
enQuireBio™
QP7346-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_058652.1 | |
Hemoglobin subunit beta-2 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
15130 | |
Research Use Only | |
Hbb-b2 | |
Recombinant Protein | |
Yeast | |
VHLTDAEKSA VSCLWAKVNP DEVGGEALGR LLVVYPWTQR YFDSFGDLSS ASAIMGNPKV KAHGKKVITA FNEGLKNLDN LKGTFASLSE LHCDKLHVDP ENFRLLGNAI VIVLGHHLGK DFTPAAQAAF QKVVAGVATA LAHKYH | |
Greater than 90% as determined by SDS-PAGE. |