Läs mer
R&D Systems™ Recombinant Mouse HGFR/c-MET Fc Chimera (NS0-expressed)
:
100ug; Carrier Free
Beskrivning
The Recombinant Mouse HGF R/c-MET Fc Chimera (NS0-expressed) is derived from NS0. The Recombinant Mouse HGF R/c-MET Fc Chimera (NS0-expressed) has been validated for the following applications: Bioactivity.
Specifikationer
Specifikationer
| Formulering | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gen-ID (Entrez) | 17295 |
| Molekylvikt (g/mol) | 96 & 32 kDa (monomer) |
| Namn | HGFR/c-MET |
| Kvantitet | 100μg |
| Källa | Mouse myeloma cell line,NS0-derived mouse HGF R/c-MET protein, (N-terminus) Mouse HGF R/c-Met (Met1-Asn929) Accession &Num; P16056 DIEGRMDP Mouse IgG2A (Glu98-Lys330) (C-terminus) |
| Förvaringskrav | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | AUTS9, c-Met, cMET, EC 2.7.10, EC 2.7.10.1, hepatocyte growth factor receptor, HGF R, HGF receptor, HGF/SF receptor, HGFR, MET, Met (c-Met), met proto-oncogene (hepatocyte growth factor receptor), met proto-oncogene tyrosine kinase, oncogene MET, Proto-oncogene c-Met, RCCP2, Scatter factor receptor, SF receptor, Tyrosine-protein kinase Met |
| Konjugera | Unconjugated |
| Rekombinant | Recombinant |
| Visa mer |