missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Histone deacetylase complex subunit SAP130 Protein
A DNA sequence encoding the Mus musculus (Mouse) Histone deacetylase complex subunit SAP130, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7805-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_766553.1 | |
Histone deacetylase complex subunit SAP130 Protein | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
269003 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Sap130 | |
Recombinant Protein | |
E. coli | |
PRKQQHVIST EEGDMMETNS TDDEKSAAKS LLVKAEKRKS PPKEYIDEEG VRYVPVRPRP PITLLRHYRN PWKAAYHHFQ RYSDVRVKEE KKAMLQEIAN QKGVSCRAQG WKVHLCAAQL LQLTNLEHDV YERLTNLQEG IIPKKKAATD DDLHRINELI QGNMQRCKLV MDQISEARDS MLKVLDHKDR VLKLLNKNGT VKKVSKLKRK EKV |