missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Hydroxyacid oxidase 1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Hydroxyacid oxidase 1, was expressed in the hosts and tags indicated.
3180.00 SEK - 38570.00 SEK
Specifications
Accession Number | NP_034533.1 |
---|---|
Gene ID (Entrez) | 15112 |
Name | Hydroxyacid oxidase 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16043611
|
enQuireBio™
QP8261-EC-100UG |
100 μg |
11125.00 SEK
100µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16063611
|
enQuireBio™
QP8261-EC-1MG |
1 mg |
38570.00 SEK
1mg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16053611
|
enQuireBio™
QP8261-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16073611
|
enQuireBio™
QP8261-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16083611
|
enQuireBio™
QP8261-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16093611
|
enQuireBio™
QP8261-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_034533.1 | |
Hydroxyacid oxidase 1 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
15112 | |
Research Use Only | |
Hao1 | |
Recombinant Protein | |
E. coli | |
MLPRLVCISD YEQHVRSVLQ KSVYDYYRSG ANDQETLADN IQAFSRWKLY PRMLRNVADI DLSTSVLGQR VSMPICVGAT AMQCMAHVDG ELATVRACQT MGTGMMLSSW ATSSIEEVAE AGPEALRWMQ LYIYKDREIS RQIVKRAEKQ GYKAIFVTVD TPYLGNRIDD VRNRFKLPPQ LRMKNFETND LAFSPKGNFG DNSGLAEYVA QAIDPSLSWD DITWLRRLTS LPIVVKGILR GDDAKEAVKH GVDGILVSNH GARQLDGVPA TIDVLPEIVE AVEGKVEVFL DGGVRKGTDV LKALALGAKA VFVGRPIIWG LAFQGEKGVQ DVLEILKEEF RLAMALSGCQ NVKVIDKTLV RKNPLAVSKI | |
Greater than 90% as determined by SDS-PAGE. |