missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse I-TAC / CXCL11 Protein

A DNA sequence encoding the Mus musculus (Mouse) I-TAC / CXCL11, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8656-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16075801

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_062367.1
I-TAC / CXCL11 Protein
Research Use Only
CXCL11
Recombinant Protein
E. coli
FLMFKQGRCL CIGPGMKAVK MAEIEKASVI YPSNGCDKVE VIVTMKAHKR QRCLDPRSKQ ARLIMQAIEK KNFLRRQ
Greater than 90% as determined by SDS-PAGE.
56066
50 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers