missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IDO1 / IDO Protein
A DNA sequence encoding the Mus musculus (Mouse) IDO1 / IDO, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9192-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001280619.1 | |
IDO1 / IDO Protein | |
Research Use Only | |
IDO1 | |
Recombinant Protein | |
Yeast | |
MALSKISPTE GSRRILEDHH IDEDVGFALP HPLVELPDAY SPWVLVARNL PVLIENGQLR EEVEKLPTLS TDGLRGHRLQ RLAHLALGYI TMAYVWNRGD DDVRKVLPRN IAVPYCELSE KLGLPPILSY ADCVLANWKK KDPNGPMTYE NMDILFSFPG GDCDKGFFLV SLLVEIAASP AIKAIPTVSS AVERQDLKAL EKALHDIATS LEKAKEIFKR MRDFVDPDTF FHVLRIYLSG WKCSSKLPEG LLYEGVWDTP KMFSGGSAGQ SSIFQSLDVL LGIKHEAGKE SPAEFLQEMR EYMPPAHRNF LFFLESAPPV REFVISRHNE DLTKAYNECV NGLVSVRKFH LAIVDTYIMK PSKKKPTDGD KSEEPSNVES RGTGGTNPMT FLRSVKDTTE KALLSWP | |
Greater than 90% as determined by SDS-PAGE. |
15930 | |
1 mg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |