missing translation for 'onlineSavingsMsg'
Läs mer

R&D Systems™ Recombinant Mouse IFNAB Protein

Produktkod. 18798356
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18798356 10 μg
18709966 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18798356 Leverantör R&D Systems™ Leverantörsnummer 10151IF010
 Begränsad artikel
Beräknad leverans: 04-12-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

Human embryonic kidney cell, HEK293-derived mouse IFNAB protein

  • The interferons (IFN) are a family of cytokines with potent antiviral, antiproliferative and immunomodulatory properties, and are classified based on their binding specificity to cell surface receptors
  • The mouse IFNAB consists of 190 amino acids (aa) including a 23 aa signal peptide and a 167 aa IFNAB main chain.
  • It was found that IFNAB plays an important role in the regulation of DNA synthesis in colong-stimulating factor (CSF)-1-stimulated macrophage, as well as mediates the inhibitory efffects of LPS and TNF alpha
TRUSTED_SUSTAINABILITY

Specifikationer

Tillträdesnummer AAA58254.1
För användning med (applikation) Bioactivity
Gen-ID (Entrez) 15974
Molekylvikt (g/mol) 19 kDa
Reningsmetod >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Kvantitet 10 μg
Källa Human embryonic kidney cell, HEK293-derived mouse IFNAB protein Cys24-Glu190
Förvaringskrav Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after opening. 3 months, -20 to -70°C under sterile conditions after opening.
Endotoxinkoncentration <0.10 EU per 1 μg of the protein by the LAL method
Konjugera Unconjugated
Rekonstitution Reconstitute at 100 μg/mL in PBS
Buffert Lyophilized from a 0.2 μm filtered solution in PBS
Produktlinje R&D Systems
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.