missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IFNG / Interferon Gamma Protein
A DNA sequence encoding the Mus musculus (Mouse) IFNG / Interferon Gamma Protein, was expressed in the hosts and tags indicated.
2430.00 SEK - 27205.00 SEK
Specifications
Accession Number | NP_032363.1 |
---|---|
Gene ID (Entrez) | 15978 |
Name | IFNG / Interferon Gamma Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16026771
|
enQuireBio™
QP8841-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16066771
|
enQuireBio™
QP8841-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16016771
|
enQuireBio™
QP8841-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16046771
|
enQuireBio™
QP8841-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16056771
|
enQuireBio™
QP8841-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16036771
|
enQuireBio™
QP8841-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_032363.1 | |
IFNG / Interferon Gamma Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
15978 | |
Research Use Only | |
IFNG | |
Recombinant Protein | |
E. coli | |
IESLESLNNY FNSSGIDVEE KSLFLDIWRN WQKDGDMKIL QSQIISFYLR LFEVLKDNQA ISNNISVIES HLITTFFSNS KAKKDAFMSI AKFEVNNPQV QRQAFNELIR VVHQLLPESS LRKRKRSRC | |
Greater than 90% as determined by SDS-PAGE. |