missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IGFBP1 Protein
A DNA sequence encoding the Mus musculus (Mouse) IGFBP1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6208-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_032367.3 | |
IGFBP1 Protein | |
Research Use Only | |
Igfbp1 | |
Recombinant Protein | |
E. coli | |
APQPWHCAPC TAERLGLCPP VPASCPEISR PAGCGCCPTC ALPMGAACGV ATARCAQGLS CRALPGEPRP LHALTRGQGA CVPEPAAPAT STLFSSQHEE AKAAVVSADE LSESPEMTEE QLLDSFHLMA PSREDQPILW NAISTYSSMR AREIADLKKW KEPCQRELYK VLERLAAAQQ KAGDEIYKFY LPNCNKNGFY HSKQCETSLD GEAGLCWCVY PWSGKKIPGS LETRGDPNCH QYFNVHN | |
Greater than 90% as determined by SDS-PAGE. |
16006 | |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |