Learn More
R&D Systems™ Recombinant Mouse IGSF2/CD101 Fc Chimera Protein, CF
Product Code. 18756016
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Product Code. Quantity Unit Size
18756016 50 μg 50µg
1 options
Product Code. 18756016 Supplier R&D Systems™ Supplier No. 3368CD050
 Restricted Item
Beräknad leverans: 04-12-2026
The Recombinant Mouse IGSF2/CD101 Fc Chimera Protein is derived from NS0
  • Recombinant Protein
  • Carrier Free (CF)
  • The carrier free version does not contain Bovine Serum Albumin (BSA)
TRUSTED_SUSTAINABILITY
Specifications
Accession Number A8E0Y8
For Use With (Application) Bioactivity
Gene ID (Entrez) 9398
Molecular Weight (g/mol) 133 kDa
Purification Method >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 50 μg
Source
Mouse myeloma cell line, NS0-derived mouse IGSF2/CD101 protein
Mouse CD101 (Gln21-Phe974) Accession # A8E0Y8IEGRMDPMouse IgG2a (Glu98-Lys330)N-terminus C-terminus
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, ≤ -20°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Reconstitution Reconstitute at 400 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS
Product Line R&D Systems