missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse IL 28A Protein

A cDNA sequence encoding the IL 28A was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10708-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15934928

  • 37420.00 SEK / 1mg
Estimated Shipment: 12-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

330496
Greater than 95% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
Ifnl2
Recombinant Protein
E. coli
DPVPRATRLP VEAKDCHIAQ FKSLSPKELQ AFKKAKDAIE KRLLEKDMRC SSHLISRAWD LKQLQVQERP KALQAEVALT LKVWENMTDS ALATILGQPL HTLSHIHSQL QTCTQLQATA EPKPPSRRLS RWLHRLQEAQ SKETPGCLED SVTSNLFRLL TRDLKCVASG DQCV
IL 28A Protein
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
The ED50 as determined by an anti-viral assay using human HepG2 cells infected with encephalomyocarditis(EMC) virus is less than 3 ng/ml, corresponding to a specific activity of > 3.3 M 105 IU/mg.
Mouse
Untagged
The Mouse IL-28A is lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4 and 5% trehalose.
Videos
SDS
Documents
Certificates
Special Offers

Special Offers