missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse IL18BP Protein

A DNA sequence encoding the Mus musculus (Mouse) IL18BP, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8880-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16056951

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_034661.1
IL18BP Protein
500 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
16068
Greater than 90% as determined by SDS-PAGE.
Research Use Only
IL18BP
Recombinant Protein
E. coli
TSAPQTTATV LTGSSKDPCS SWSPAVPTKQ YPALDVIWPE KEVPLNGTLT LSCTACSRFP YFSILYWLGN GSFIEHLPGR LKEGHTSREH RNTSTWLHRA LVLEELSPTL RSTNFSCLFV DPGQVAQYHI ILAQLWDGLK TAPSPSQETL SSHSPVSRSA GPGVA
Videos
SDS
Documents