missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL1RL2 / IL-1Rrp2 Protein
A DNA sequence encoding the Mus musculus (Mouse) IL1RL2 / IL-1Rrp2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8137-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_573456.1 | |
IL1RL2 / IL-1Rrp2 Protein | |
Research Use Only | |
Il1rl2 | |
Recombinant Protein | |
E. coli | |
DTCEDIFMHN VIISEGQPFP FNCTYPPETN GAVNLTWYKT PSKSPVSNNR HLRVHQDQTW ILFLPLTLED SGIYQCVIRN AHNCYQIAVN LTVLKNHWCD SSMEGSPVNS PDVYQQILPI GKSGSLNCHL YFPESCALDS IKWYKGCEEI KAGKKYSPSG AKLLVNNVAV EDGGSYACSA RLTHLGRHFT IRNYIAVNTK EVEYGRRIPN ITYPKNNSIE VPLGSTLIVN CNITDTKENT NLRCWRVNNT LVDDYYKDSK RIQEGIETNV SLRDQIRYTV NITFLKVKME DYGRPFTCHA GVSAAYIILI YPVPDFR | |
Greater than 90% as determined by SDS-PAGE. |
107527 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |