missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse ITGB1 / Integrin beta-1 / CD29 Protein
A DNA sequence encoding the Mus musculus (Mouse) ITGB1 / Integrin beta-1 / CD29, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6241-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_034708.1 | |
ITGB1 / Integrin beta-1 / CD29 Protein | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
16412 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Itgb1 | |
Recombinant Protein | |
E. coli | |
QTDKNRCLKA NAKSCGECIQ AGPNCGWCTN TTFLQEGMPT SARCDDLEAL KKKGCQPSDI ENPRGSQTIK KNKNVTNRSK GMAEKLRPED ITQIQPQQLL LKLRSGEPQK FTLKFKRAED YPIDLYYLMD LSYSMKDDLE NVKSLGTDLM NEMRRITSDF RIGFGSFVEK TVMPYISTTP AKLRNPCTSE QNCTSPFSYK NVLSLTDRGE FFNELVGQQR ISGNLDSPEG GFDAIMQVAV CGSLIGWRNV TRLLVFSTDA GFHFAGDGKL GGIVLPNDGQ CHLENNVYTM SHYYDYPSIA HLVQKLSENN IQTIFAVTEE FQPVYKELKN LIPKSAVGTL SGNSSNVIQL IIDAYNSLSS EVILENSKLP DGVTINYKSY CKNGVNGTGE NGRKCSNISI GDEVQFEISI TANKCPNKES ETIKIKPLGF TEEVEVVLQF ICKCNCQSHG IPASPKCHEG NGTFECGACR CNEGRVGRHC ECSTDEVNSE DMDAYCRKEN SSEICSNNGE CVCGQCVCRK RDNTNEIYSG KFCECDNFNC DRSNGLICGG NGVCRCRVCE CYPNYTGSAC DCSLDTGPCL ASNGQICNGR GICECGACKC TDPKFQGPTC ETCQTCLGVC AEHKECVQCR AFNKGEKKDT CAQECSHFNL TKVESREKLP QPVQVDPVTH CKEKDIDDCW FYFTYSVNGN NEAIVHVVET PDCPTGPD |