missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Kinesin-like protein KIF1A Protein
A DNA sequence encoding the Mus musculus (Mouse) Kinesin-like protein KIF1A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6262-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
Kinesin-like protein KIF1A Protein | |
Research Use Only | |
Kif1a | |
Recombinant Protein | |
E. coli | |
MAGASVKVAV RVRPFNSREM SRDSKCIIQM SGSTTTIVNP KQPKETPKSF SFDYSYWSHT SPEDINYASQ KQVYRDIGEE MLQHAFEGYN VCIFAYGQTG AGKSYTMMGK QEKDQQGIIP QLCEDLFSRI NDTTNDNMSY SVEVSYMEIY CERVRDLLNP KNKGNLRVRE HPLLGPYVED LSKLAVTSYN DIQDLMDSGN KPRTVAATNM NETSSRSHAV FNIIFTQKRH DAETNITTEK VSKISLVDLA GSERADSTGA KGTRLKEGAN INKSLTTLGK VISALAEMDS GPNKNKKKKK TDFIPYRDSV LTWLLRENLG GNSRTAMVAA LSPADINYDE TLSTLRYADR AKQIRCNAII N | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |