missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Kinesin-like protein KIF1A Protein

A DNA sequence encoding the Mus musculus (Mouse) Kinesin-like protein KIF1A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6262-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16052010

  • 38570.00 SEK / 1mg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Kinesin-like protein KIF1A Protein
Research Use Only
Kif1a
Recombinant Protein
E. coli
MAGASVKVAV RVRPFNSREM SRDSKCIIQM SGSTTTIVNP KQPKETPKSF SFDYSYWSHT SPEDINYASQ KQVYRDIGEE MLQHAFEGYN VCIFAYGQTG AGKSYTMMGK QEKDQQGIIP QLCEDLFSRI NDTTNDNMSY SVEVSYMEIY CERVRDLLNP KNKGNLRVRE HPLLGPYVED LSKLAVTSYN DIQDLMDSGN KPRTVAATNM NETSSRSHAV FNIIFTQKRH DAETNITTEK VSKISLVDLA GSERADSTGA KGTRLKEGAN INKSLTTLGK VISALAEMDS GPNKNKKKKK TDFIPYRDSV LTWLLRENLG GNSRTAMVAA LSPADINYDE TLSTLRYADR AKQIRCNAII N
Greater than 90% as determined by SDS-PAGE.
1 mg
Not determined.
Partial (see sequence information for more details).
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers