missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Legumain / LGMN Protein
A DNA sequence encoding the Mus musculus (Mouse) Legumain/ LGMN, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6302-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_035305.1 | |
Legumain / LGMN Protein | |
Research Use Only | |
Lgmn | |
Recombinant Protein | |
Yeast | |
VPVGVDDPED GGKHWVVIVA GSNGWYNYRH QADACHAYQI IHRNGIPDEQ IIVMMYDDIA NSEENPTPGV VINRPNGTDV YKGVLKDYTG EDVTPENFLA VLRGDAEAVK GKGSGKVLKS GPRDHVFIYF TDHGATGILV FPNDDLHVKD LNKTIRYMYE HKMYQKMVFY IEACESGSMM NHLPDDINVY ATTAANPKES SYACYYDEER GTYLGDWYSV NWMEDSDVED LTKETLHKQY HLVKSHTNTS HVMQYGNKSI STMKVMQFQG MKHRASSPIS LPPVTHLDLT PSPDVPLTIL KRKLLRTN | |
Greater than 90% as determined by SDS-PAGE. |
19141 | |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |