missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Major urinary protein 1

A DNA sequence encoding the Mus musculus (Mouse) Major urinary protein 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7038-ye-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16047150

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

Major urinary protein 1
Research Use Only
Mup2
Recombinant Protein
Yeast
EEASSTGRNF NVEKINGEWH TIILASDKRE KIEDNGNFRL FLEQIHVLEK SLVLKFHTVR DEECSELSMV ADKTEKAGEY SVTYDGFNTF TIPKTDYDNF LMAHLINEKD GETFQLMGLY GREPDLSSDI KERFAKLCEE HGILRENIID LSNANRCLQA RE
Greater than 90% as determined by SDS-PAGE.
200 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers