missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse MCL1 homolog Protein
A DNA sequence encoding the Mus musculus (Mouse) MCL1 homolog, was expressed in the hosts and tags indicated.
2430.00 SEK - 38570.00 SEK
Specifications
Accession Number | NP_032588.1 |
---|---|
Gene ID (Entrez) | 17210 |
Name | MCL1 homolog Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16082650
|
enQuireBio™
QP6348-EC-100UG |
100 μg |
8695.00 SEK
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16002660
|
enQuireBio™
QP6348-EC-1MG |
1 mg |
38570.00 SEK
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16092650
|
enQuireBio™
QP6348-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16012660
|
enQuireBio™
QP6348-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16022660
|
enQuireBio™
QP6348-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16032660
|
enQuireBio™
QP6348-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_032588.1 | |
MCL1 homolog Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
17210 | |
Research Use Only | |
Mcl1 | |
Recombinant Protein | |
E. coli | |
MFGLRRNAVI GLNLYCGGAS LGAGGGSPAG ARLVAEEAKA RREGGGEAAL LPGARVVARP PPVGAEDPDV TASAERRLHK SPGLLAVPPE EMAASAAAAI VSPEEELDGC EPEAIGKRPA VLPLLERVSE AAKSSGADGS LPSTPPPPEE EEDDLYRQSL EIISRYLREQ ATGSKDSKPL GEAGAAGRRA LETLRRVGDG VQRNHETAFQ GMLRKLDIKN EGDVKSFSRV MVHVFKDGVT NWGRIVTLIS FGAFVAKHLK SVNQESFIEP LAETITDVLV RTKRDWLVKQ RGWDGFVEFF HVQDLEGG | |
Greater than 90% as determined by SDS-PAGE. |