missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Nmes1 Protein

A DNA sequence encoding the Mus musculus (Mouse) Nmes1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7786-ma-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16060791

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001004174.1
Nmes1 Protein
Research Use Only
Nmes1
Recombinant Protein
Mammal
MGVFQILMKN KELIPLAFFI SVAATGATSF ALYALKKTDV VIDRKRNPEP WEMVDPTQPQ KLITINQQWK PVEELQKVRR ATR
Greater than 90% as determined by SDS-PAGE.
433470
50 μg
Not determined.
Full Length
Mouse
Flag
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers