missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse NPM1 / Nucleophosmin Protein
A DNA sequence encoding the Mus musculus (Mouse) NPM1 / Nucleophosmin, was expressed in the hosts and tags indicated.
2515.00 SEK - 38570.00 SEK
Specifications
Accession Number | NP_032748.1 |
---|---|
Gene ID (Entrez) | 18148 |
Name | NPM1 / Nucleophosmin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16040491
|
enQuireBio™
QP7731-EC-100UG |
100 μg |
8695.00 SEK
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16060491
|
enQuireBio™
QP7731-EC-1MG |
1 mg |
38570.00 SEK
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16050491
|
enQuireBio™
QP7731-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16070491
|
enQuireBio™
QP7731-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16080491
|
enQuireBio™
QP7731-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16090491
|
enQuireBio™
QP7731-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Spezifikation
NP_032748.1 | |
NPM1 / Nucleophosmin Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
18148 | |
Research Use Only | |
Npm1 | |
Recombinant Protein | |
E. coli | |
MEDSMDMDMS PLRPQNYLFG CELKADKDYH FKVDNDENEH QLSLRTVSLG AGAKDELHIV EAEAMNYEGS PIKVTLATLK MSVQPTVSLG GFEITPPVVL RLKCGSGPVH ISGQHLVAVE EDAESEDEDE EDVKLLGMSG KRSAPGGGNK VPQKKVKLDE DDEDDDEDDE DDEDDDDDDF DEEETEEKVP VKKSVRDTPA KNAQKSNQNG KDLKPSTPRS KGQESFKKQE KTPKTPKGPS SVEDIKAKMQ ASIEKGGSLP KVEAKFINYV KNCFRMTDQE AIQDLWQWRK SL | |
Greater than 90% as determined by SDS-PAGE. |