missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Nuclear receptor ROR-gamma Protein
A DNA sequence encoding the Mus musculus (Mouse) Nuclear receptor ROR-gamma, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9374-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001280663.1 | |
Nuclear receptor ROR-gamma Protein | |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
19885 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
RORC | |
Recombinant Protein | |
Yeast | |
MDRAPQRHHR TSRELLAAKK THTSQIEVIP CKICGDKSSG IHYGVITCEG CKGFFRRSQQ CNVAYSCTRQ QNCPIDRTSR NRCQHCRLQK CLALGMSRDA VKFGRMSKKQ RDSLHAEVQK QLQQQQQQEQ VAKTPPAGSR GADTLTYTLG LSDGQLPLGA SPDLPEASAC PPGLLRASGS GPPYSNTLAK TEVQGASCHL EYSPERGKAE GRDSIYSTDG QLTLGRCGLR FEETRHPELG EPEQGPDSHC IPSFCSAPEV PYASLTDIEY LVQNVCKSFR ETCQLRLEDL LRQRTNLFSR EEVTSYQRKS MWEMWERCAH HLTEAIQYVV EFAKRLSGFM ELCQNDQIIL LTAGAMEVVL VRMCRAYNAN NHTVFFEGKY GGVELFRALG CSELISSIFD FSHFLSALCF SEDEIALYTA LVLINANRPG LQEKRRVEHL QYNLELAFHH HLCKTHRQGL LAKLPPKGKL RSLCSQHVEK LQIFQHLHPI VVQAAFPPLY KELFSTDVES PEGLSK |