missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse OTC Protein

A DNA sequence encoding the Mus musculus (Mouse) OTC, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6457-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16003440

  • 38570.00 SEK / 1mg
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_032795.1
OTC Protein
Research Use Only
OTC
Recombinant Protein
E. coli
SQVQLKGRDL LTLKNFTGEE IQYMLWLSAD LKFRIKQKGE YLPLLQGKSL GMIFEKRSTR TRLSTETGFA LLGGHPSFLT TQDIHLGVNE SLTDTARVLS SMTDAVLARV YKQSDLDTLA KEASIPIVNG LSDLYHPIQI LADYLTLQEH YGSLKGLTLS WIGDGNNILH SIMMSAAKFG MHLQAATPKG YEPDPNIVKL AEQYAKENGT KLSMTNDPLE AARGGNVLIT DTWISMGQED EKKKRLQAFQ GYQVTMKTAK VAASDWTFLH CLPRKPEEVD DEVFYSPRSL VFPEAENRKW TIMAVMVSLL TDYSPVLQKP KF
Greater than 90% as determined by SDS-PAGE.
18416
1 mg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers