missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Proto-oncogene Wnt-3 Protein

A DNA sequence encoding the Mus musculus (Mouse) Proto-oncogene Wnt-3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9467-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16098361

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_033547.1
Proto-oncogene Wnt-3 Protein
Research Use Only
Wnt3
Recombinant Protein
Yeast
GYPIWWSLAL GQQYTSLASQ PLLCGSIPGL VPKQLRFCRN YIEIMPSVAE GVKLGIQECQ HQFRGRRWNC TTIDDSLAIF GPVLDKATRE SAFVHAIASA GVAFAVTRSC AEGTSTICGC DSHHKGPPGE GWKWGGCSED ADFGVLVSRE FADARENRPD ARSAMNKHNN EAGRTTILDH MHLKCKCHGL SGSCEVKTCW WAQPDFRAIG DFLKDKYDSA SEMVVEKHRE SRGWVETLRA KYALFKPPTE RDLVYYENSP NFCEPNPETG SFGTRDRTCN VTSHGIDGCD LLCCGRGHNT RTEKRKEKCH CVF
Greater than 90% as determined by SDS-PAGE.
22415
1 mg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers