missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Pyridoxal phosphate phosphatase Protein
A DNA sequence encoding the Mus musculus (Mouse) Pyridoxal phosphate phosphatase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6482-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_064667.2 | |
Pyridoxal phosphate phosphatase Protein | |
Research Use Only | |
Pdxp | |
Recombinant Protein | |
E. coli | |
MARCERLRGA ALRDVLGQAQ GVLFDCDGVL WNGERIVPGA PELLQRLARA GKNTLFVSNN SRRARPELAL RFARLGFAGL RAEQLFSSAL CAARLLRQRL SGPPDASGAV FVLGGEGLRA ELRAAGLRLA GDPGEDPRVR AVLVGYDEQF SFSRLTEACA HLRDPDCLLV ATDRDPWHPL SDGSRTPGTG SLAAAVETAS GRQALVVGKP SPYMFQCITE DFSVDPARTL MVGDRLETDI LFGHRCGMTT VLTLTGVSSL EEAQAYLTAG QRDLVPHYYV ESIADLMEGL ED | |
Greater than 90% as determined by SDS-PAGE. |
57028 | |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |