missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Ras-related protein Rab-10 Protein
A DNA sequence encoding the Mus musculus (Mouse) Ras-related protein Rab-10, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6565-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_057885.1 | |
Ras-related protein Rab-10 Protein | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
19325 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Rab10 | |
Recombinant Protein | |
E. coli | |
MAKKTYDLLF KLLLIGDSGV GKTCVLFRFS DDAFNTTFIS TIGIDFKIKT VELQGKKIKL QIWDTAGQER FHTITTSYYR GAMGIMLVYD ITNGKSFENI SKWLRNIDEH ANEDVERMLL GNKCDMDDKR VVPKGKGEQI AREHGIRFFE TSAKANINIE KAFLTLAEDI LRKTPVKEPN SENVDISSGG GVTGWKSKCC |