missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse SAMHD1 Protein
A DNA sequence encoding the Mus musculus (Mouse) SAMHD1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6650-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001132992.1 | |
SAMHD1 Protein | |
Research Use Only | |
SAMHD1 | |
Recombinant Protein | |
E. coli | |
DIMITDAFLK ADPYVEITGT AGKKFRISTA IDDMEAFTKL TDNIFLEVLH STDPQLSEAQ SILRNIECRN LYKYLGETQP KREKIRKEEY ERLPQEVAKA KPEKAPDVEL KAEDFIVDVI NVDYGMEDKN PIDRVHFYCK SNSKQAVRIN KEQVSQLLPE KFAEQLIRVY CKKKDGKSLD AAGKHFVQWC ALRDFTKPQD GDIIAPLITP LKWNNKTSSC LQEVSKVKTC LK | |
Greater than 90% as determined by SDS-PAGE. |
56045 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |