missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Scrapie-responsive protein 1
A DNA sequence encoding the Mus musculus (Mouse) Scrapie-responsive protein 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7487-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_033162.1 | |
Scrapie-responsive protein 1 | |
Research Use Only | |
SCRG1 | |
Recombinant Protein | |
Yeast | |
MPSSRLSCYR KLLKDRNCHN LPEGRADLKL IDANVQHHFW DGKGCEMICY CNFSELLCCP KDVFFGPKIS FVIPCNNH | |
Greater than 90% as determined by SDS-PAGE. |
20284 | |
1 mg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |