missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse SDF2 Protein
A DNA sequence encoding the Mus musculus (Mouse) SDF2, was expressed in the hosts and tags indicated.
2430.00 SEK - 38570.00 SEK
Specifications
Accession Number | NP_033169.3 |
---|---|
Gene ID (Entrez) | 20316 |
Name | SDF2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16044870
|
enQuireBio™
QP6658-EC-100UG |
100 μg |
8695.00 SEK
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16064870
|
enQuireBio™
QP6658-EC-1MG |
1 mg |
38570.00 SEK
1mg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16054870
|
enQuireBio™
QP6658-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16074870
|
enQuireBio™
QP6658-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16084870
|
enQuireBio™
QP6658-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16094870
|
enQuireBio™
QP6658-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_033169.3 | |
SDF2 Protein | |
Not determined. | |
Full Length of Mature Protein | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
20316 | |
Research Use Only | |
SDF2 | |
Recombinant Protein | |
E. coli | |
SNMAVVTCGS VVKLLNTRHN VRLHSHDVRY GSGSGQQSVT GVTSVDDSNS YWRIRGKTAT VCERGTPIKC GQPIRLTHIN TGRNLHSHHF TSPLSGNQEV SAFGEEGEGD YLDDWTVLCN GPYWVRDGEV RFKHSSTDVL LSVTGEQYGR PISGQKEVHG MAQPSQNNYW KAMEGIFMKP SELLRAEVHH AEL | |
Greater than 80% as determined by SDS-PAGE. |