missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Secretoglobin family 3A member 2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Secretoglobin family 3A member 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7929-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001276572.1 | |
Secretoglobin family 3A member 2 Protein | |
Research Use Only | |
Scgb3a2 | |
Recombinant Protein | |
Yeast | |
LLINRLPVVD KLPVPLDDII PSFDPLKMLL KTLGISVEHL VTGLKKCVDE LGPEASEAVK KLLVIIICSY FPGRSLCYVN NLPSFVSVLF LPMICAYPRD SKKQTFAFIE RVFEQSKL | |
Greater than 80% as determined by SDS-PAGE. |
117158 | |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |