missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Serpina3n Protein
A DNA sequence encoding the Mus musculus (Mouse) Serpina3n, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7862-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
Serpina3n Protein | |
1 mg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Serpina3n | |
Recombinant Protein | |
Yeast | |
FPDGTLGMDA AVQEDHDNGT QLDSLTLASI NTDFAFSLYK ELVLKNPDKN IVFSPLSISA ALAVMSLGAK GNTLEEILEG LKFNLTETSE ADIHQGFGHL LQRLNQPKDQ VQISTGSALF IEKRQQILTE FQEKAKTLYQ AEAFTADFQQ PRQAKKLIND YVRKQTQGMI KELVSDLDKR TLMVLVNYIY FKAKWKVPFD PLDTFKSEFY AGKRRPVIVP MMSMEDLTTP YFRDEELSCT VVELKYTGNA SALFILPDQG RMQQVEASLQ PETLRKWKNS LKPRMIDELH LPKFSISTDY SLEDVLSKLG IREVFSTQAD LSAITGTKDL RVSQVVHKAV LDVAETGTEA AAATGVKFVP MSAKLYPLTV YFNRPFLIMI FDTETEIAPF IAKIANPK |