missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Serum amyloid A-3 Protein

A DNA sequence encoding the Mus musculus (Mouse) Serum amyloid A-3 protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7359-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16088430

  • 24845.00 SEK / 500µg
Estimated Shipment: 04-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_035445.1
Serum amyloid A-3 Protein
Research Use Only
Saa3
Recombinant Protein
Yeast
RWVQFMKEAG QGSRDMWRAY SDMKKANWKN SDKYFHARGN YDAARRGPGG AWAAKVISDA REAVQKFTGH GAEDSRADQF ANEWGRSGKD PNHFRPAGLP KRY
Greater than 80% as determined by SDS-PAGE.
20210
500 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers