missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse SIGIRR / TIR8 Protein
A DNA sequence encoding the Mus musculus (Mouse) SIGIRR / TIR8, was expressed in the hosts and tags indicated.
3180.00 SEK - 38570.00 SEK
Specifications
Accession Number | NP_075546.2 |
---|---|
Gene ID (Entrez) | 24058 |
Name | SIGIRR / TIR8 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16063351
|
enQuireBio™
QP8219-EC-100UG |
100 μg |
11125.00 SEK
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16083351
|
enQuireBio™
QP8219-EC-1MG |
1 mg |
38570.00 SEK
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16073351
|
enQuireBio™
QP8219-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16093351
|
enQuireBio™
QP8219-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16003361
|
enQuireBio™
QP8219-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16013361
|
enQuireBio™
QP8219-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_075546.2 | |
SIGIRR / TIR8 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
24058 | |
Research Use Only | |
Sigirr | |
Recombinant Protein | |
E. coli | |
MAGVCDMAPN FLSPSEDQAL GLALGREVAL NCTAWVFSRP QCPQPSVQWL KDGLALGNGS HFSLHEDFWV SANFSEIVSS VLVLNLTNAE DYGTFTCSVW NVSSHSFTLW RAGPAGH | |
Greater than 90% as determined by SDS-PAGE. |